Learn More
Biomatik Corporation Rat Abcb1 (Multidrug resistance protein 1) ELISA Kit
Supplier:  Biomatik Corporation EKF58047
Encompass_Preferred
Rat Abcb1 (Multidrug resistance protein 1) ELISA Kit for detecting Abcb1 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.313-20ng/ml. Sensitivity: 0.188ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P43245. Target Synonyms: Abcb1; ABC20; ABCB1; ABCB1B; CD243; CLCS; GP170; IBD13; PGY1; ABC20; ABCB1; ABCB1B; ATP-binding cassette sub-family B member 1; ATP-binding cassette; sub-family B; MDR; TAP; member 1; CD243; CD243 antigen; CLCS; colchicin sensitivity; doxorubicin resistance; EC 3.6.3; EC 3.6.3.44; GP170; IBD13; MDR1MGC163296; multidrug resistance protein 1; P-glycoprotein 1; P-gp; PGY1; PGY1P-GP. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5504