Learn More
Biomatik Corporation Mouse IFABP/FABP2 (Intestinal Fatty Acid Binding Protein) ELISA Kit
Supplier:  Biomatik Corporation EKF58075
Encompass_Preferred
Mouse IFABP/FABP2 (Intestinal Fatty Acid Binding Protein) ELISA Kit for detecting IFABP; FABP2 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P55050. Target Synonyms: IFABP; FABP2; FABPI; I-FABP; I-FABP; Intestinal FABP; FABPI; fatty acid binding protein 2; intestinal; Fatty acid-binding protein 2; fatty acid-binding protein; intestinal; I-FABP; Intestinal-type fatty acid-binding protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5522