Learn More
Biomatik Corporation Rat FXI (Coagulation Factor XI) ELISA Kit
Supplier:  Biomatik Corporation EKF58081
Encompass_Preferred
Rat FXI (Coagulation Factor XI) ELISA Kit for detecting FXI in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: A0A0G2K4I9. Target Synonyms: FXI. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5527