Learn More
Biomatik Corporation Human 26S-PSM (26S Proteasome) ELISA Kit
Supplier:  Biomatik Corporation EKF58101
Encompass_Preferred
Human 26S-PSM (26S Proteasome) ELISA Kit for detecting 26S-PSM in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.313-20ng/ml. Sensitivity: 0.188ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P20618; P25788. Target Synonyms: 26S-PSM; 26S Proteasome; 26SPSM; 26-SPSM. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5542