Learn More
Biomatik Corporation Mouse SRC1 (Steroid receptor coactivator 1) ELISA Kit
Supplier:  Biomatik Corporation EKF58108
Encompass_Preferred
Mouse SRC1 (Steroid receptor coactivator 1) ELISA Kit for detecting SRC1 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.625-1000pg/ml. Sensitivity: 9.375pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P70365. Target Synonyms: bHLHe42; bHLHe74; F SRC 1; KAT13A; NCoA 1; NCOA1; nuclear receptor coactivator 1; Protein Hin 2; RIP160; SRC 1; SRC1; Steroid receptor coactivator 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5549