Learn More
Biomatik Corporation Rat GHRL (Ghrelin) ELISA Kit
Supplier:  Biomatik Corporation EKF58122
Encompass_Preferred
Rat GHRL (Ghrelin) ELISA Kit for detecting GHRL in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 7.813-500pg/ml. Sensitivity: 4.688pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q9QYH7. Target Synonyms: Ghrl; ghrelin; ghrelin; obestatin preprohormone; ghrelin; obestatin prepropeptide; Growth hormone secretagogue; Growth hormone-releasing peptide; Motilin-related peptide; MTLRPgrowth hormone secretagogue receptor ligand; obestatin; Protein M46. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5560