Learn More
Biomatik Corporation Mouse Mdk (Midkine) ELISA Kit
Supplier:  Biomatik Corporation EKF58140
Encompass_Preferred
Mouse Mdk (Midkine) ELISA Kit for detecting Mdk in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 78.125-5000pg/ml. Sensitivity: 46.875pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P12025. Target Synonyms: Mdk; MK; Midkine; MDK; ARAP; MEK; MK1; NEGF2; Amphiregulin-associated protein; Midgestation and kidney protein; midkine; midkine; neurite growth-promoting factor 2; Neurite outgrowth-promoting factor 2; Neurite outgrowth-promoting protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5572