Learn More
Biomatik Corporation Mouse FOXO3 (Forkhead box protein O3) ELISA Kit
Supplier:  Biomatik Corporation EKF58143
Encompass_Preferred
Mouse FOXO3 (Forkhead box protein O3) ELISA Kit for detecting FOXO3 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q9WVH4. Target Synonyms: FOXO3; Forkhead box protein O3; AF6q21; FKHRL1; FKHRL1P2; FOXO3A; forkhead box O3; Forkhead in rhabdomyosarcoma-like 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5574