Learn More
Biomatik Corporation Human CCL17 (C-C motif chemokine 17) ELISA Kit
Supplier:  Biomatik Corporation EKF58158
Encompass_Preferred
Human CCL17 (C-C motif chemokine 17) ELISA Kit for detecting CCL17 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 31.25-2000pg/ml. Sensitivity: 18.75pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q92583. Target Synonyms: CCL17; C-C motif chemokine 17; TARC; ABCD-2; SCYA17; Thymus Activation Regulated Chemokine. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5585