Learn More
Biomatik Corporation Rat NE (Neutrophil Elastase) ELISA Kit
Supplier:  Biomatik Corporation EKF58182
Encompass_Preferred
Rat NE (Neutrophil Elastase) ELISA Kit for detecting NE in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 1.563-100ng/ml. Sensitivity: 0.938ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P00774. Target Synonyms: NE; ELANE; PMN elastase; Medullasin; Bone marrow serine protease; Elastase-2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5601