Learn More
Biomatik Corporation Rat IL-1α (Interleukin 1 Alpha) ELISA Kit
Supplier:  Biomatik Corporation EKF58189
Encompass_Preferred
Rat IL-1α (Interleukin 1 Alpha) ELISA Kit for detecting IL-1α in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 15.625-1000pg/ml. Sensitivity: 9.375pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P16598. Target Synonyms: IL-1α; IL-1alpha; IL1A; IL1-A; IL1; IL1F1; Preinterleukin 1 Alpha; Hematopoietin-1; Pro-Interleukin-1-Alpha. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5606