Learn More
Biomatik Corporation Rat TWEAK (Tumour Necrosis Factor Related Weak Inducer of Apoptosis) ELISA Kit
Supplier:  Biomatik Corporation EKF58194
Encompass_Preferred
Rat TWEAK (Tumour Necrosis Factor Related Weak Inducer of Apoptosis) ELISA Kit for detecting TWEAK in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 15.625-1000pg/ml. Sensitivity: 9.375pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q6J1A8. Target Synonyms: TWEAK; TNFSF12; APO3 ligand; APO3LMGC20669; DR3LGTWEAKAPO3; DR3 ligand; MGC129581; TNF-related weak inducer of apoptosis; tumor necrosis factor; ligand superfamily; member 12; tumor necrosis factor ligand superfamily member 12. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5611