Learn More
Biomatik Corporation Rat Apob (Apolipoprotein B) ELISA Kit
Supplier:  Biomatik Corporation EKF58199
Encompass_Preferred
Rat Apob (Apolipoprotein B) ELISA Kit for detecting Apob in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 3.125-200ng/ml. Sensitivity: 1.875ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q7TMA5. Target Synonyms: Apob; Apolipoprotein B; APOB; FLDB; apoB-48; apolipoprotein B48; FLDB; LDLCQ4. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5613