Learn More
Biomatik Corporation Mouse CRP (C-reactive protein) ELISA Kit
Supplier:  Biomatik Corporation EKF58208
Encompass_Preferred
Mouse CRP (C-reactive protein) ELISA Kit for detecting CRP in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.391-25ng/ml. Sensitivity: 0.234ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P14847. Target Synonyms: CRP; C-Reactive Protein; PTX1; Pentraxin 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5620