Learn More
Biomatik Corporation Rat GDH (Glutamate Dehydrogenase) ELISA Kit
Supplier:  Biomatik Corporation EKF58237
Encompass_Preferred
Rat GDH (Glutamate Dehydrogenase) ELISA Kit for detecting GDH in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.625-40ng/ml. Sensitivity: 0.375ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P10860. Target Synonyms: GDH; GLDH; Glutamate dehydrogenase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5639