Learn More
Biomatik Corporation Mouse Rorc (Nuclear receptor ROR-gamma) ELISA Kit
Supplier:  Biomatik Corporation EKF58244
Encompass_Preferred
Mouse Rorc (Nuclear receptor ROR-gamma) ELISA Kit for detecting Rorc in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 46.875-3000pg/ml. Sensitivity: 28.125pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P51450. Target Synonyms: RORC; ROR gamma; NR1F3; RZRG; MGC129539; nuclear receptor ROR-gamma; Nuclear receptor RZR-gamma; Nuclear receptor subfamily 1 group F member 3; RAR-related orphan receptor C; RAR-related orphan receptor gamma; retinoic acid-binding receptor gamma; retinoid-related orphan receptor gamma; Retinoid-related orphan receptor-gamma; RORGMGC129539; RZRGRZR-GAMMA; TOR. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5645