Learn More
Biomatik Corporation Mouse NRG-4 (Neuregulin 4) ELISA Kit
Supplier:  Biomatik Corporation EKF58249
Encompass_Preferred
Mouse NRG-4 (Neuregulin 4) ELISA Kit for detecting NRG-4 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.313-20ng/ml. Sensitivity: 0.188ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q9WTX4. Target Synonyms: NRG-4. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5649