Learn More
Biomatik Corporation Human Indian hedgehog (IHH) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC34122
Encompass_Preferred
The Human Indian hedgehog (IHH) ELISA Kit allows for quantitative determination indian hedgehog homolog (Drosophila) in serum, plasma and tissue homogenates. Species Reactivity: Human. Detection Range: 1.56 pg/mL-100 pg/mL. Sensitivity: 0.39 pg/mL. Format: 96-Well Plate. Assay Type: ELISA. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Q14623. Restriction: For Research Use Only.
Catalog No. 50-149-7991