Learn More
Biomatik Corporation Mouse Rxrb (Retinoic acid receptor RXR-beta) ELISA Kit
Supplier:  Biomatik Corporation EKF58261
Encompass_Preferred
Mouse Rxrb (Retinoic acid receptor RXR-beta) ELISA Kit for detecting Rxrb in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.313-20ng/ml. Sensitivity: 0.188ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P28704. Target Synonyms: Rxrb; RXRB; NR2B2; Retinoic acid receptor RXR-beta; Nuclear receptor subfamily 2 group B member 2; Retinoid X receptor beta. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5659