Learn More
Biomatik Corporation Mouse LZM (Lysozyme) ELISA Kit
Supplier:  Biomatik Corporation EKF58269
Encompass_Preferred
Mouse LZM (Lysozyme) ELISA Kit for detecting LZM in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 31.25-2000ng/ml. Sensitivity: 18.75ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P17897. Target Synonyms: LZM; lysozyme; lysozyme; renal amyloidosis1; 4-beta-N-acetylmuramidase C; lysozyme C; LYZ; renal amyloidosis. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5664