Learn More
Biomatik Corporation Human FNDC5 (Fibronectin type III domain-containing protein 5) ELISA Kit
Supplier:  Biomatik Corporation EKF58308
Encompass_Preferred
Human FNDC5 (Fibronectin type III domain-containing protein 5) ELISA Kit for detecting FNDC5 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 78.125-5000pg/ml. Sensitivity: 46.875pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q8NAU1. Target Synonyms: FNDC5; Fibronectin type III domain-containing protein 5; Fibronectin type III repeat-containing protein 2; Irisin; FRCP2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5688