Learn More
Biomatik Corporation Rat ALT (Alanine Transaminase) ELISA Kit
Supplier:  Biomatik Corporation EKF58317
Encompass_Preferred
Rat ALT (Alanine Transaminase) ELISA Kit for detecting ALT in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 1.563-100ng/ml. Sensitivity: 0.938ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: A0A0G2JV84; A0A8I6ARL0; Q7TP13. Target Synonyms: ALT; Alanine Transaminase; ALAT; SGPT; alanine aminotransferase; serum glutamic pyruvic transaminase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5694