Learn More
Biomatik Corporation Mouse CRT (Calreticulin) ELISA Kit
Supplier:  Biomatik Corporation EKF58320
Encompass_Preferred
Mouse CRT (Calreticulin) ELISA Kit for detecting CRT in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P14211. Target Synonyms: CRT; Autoantigen Ro; CALR; cC1qR; SSA; Vasostatin; autoantigen Ro; calregulin; Calregulin; calreticulin; CRP55; CRTC; Endoplasmic reticulum resident protein 60; ERp60; FLJ26680; grp60; RO; Sicca syndrome antigen A; autoantigen Ro; calreticulin; SSAHACBP. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5697