Learn More
Biomatik Corporation Mouse BMI-1 (Polycomb complex protein BMI-1) ELISA Kit
Supplier:  Biomatik Corporation EKF58325
Encompass_Preferred
Mouse BMI-1 (Polycomb complex protein BMI-1) ELISA Kit for detecting BMI-1 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 7.813-500pg/ml. Sensitivity: 4.688pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P25916. Target Synonyms: B lymphoma Mo-MLV insertion region 1 homolog; flvi-2; bmi-1; murine leukemia viral; bmi-1 oncogene homolog; oncogene BMI-1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5701