Learn More
Biomatik Corporation Mouse NK1R (Neurokinin-1 receptor) ELISA Kit
Supplier:  Biomatik Corporation EKF58327
Encompass_Preferred
Mouse NK1R (Neurokinin-1 receptor) ELISA Kit for detecting NK1R in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 46.875-3000pg/ml. Sensitivity: 28.125pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P30548. Target Synonyms: Neurokinin-1 receptor; NK 1 receptor; NK 1R; NK1R; NKIR; SPR; Substance P receptor; TAC1R; tachykinin receptor 1; TACR1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5703