Learn More
Biomatik Corporation Rat Esr1 (Estrogen receptor) ELISA Kit
Supplier:  Biomatik Corporation EKF58346
Encompass_Preferred
Rat Esr1 (Estrogen receptor) ELISA Kit for detecting Esr1 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.313-20ng/ml. Sensitivity: 0.188ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P06211. Target Synonyms: Esr1; ER alpha; NR3A1; ER; Era; ER-alpha; Estradiol receptor; estrogen receptor; estrogen receptor 1; Nuclear receptor subfamily 3 group A member 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5715