Learn More
Biomatik Corporation Human PLG (Plasminogen) ELISA Kit
Supplier:  Biomatik Corporation EKF58357
Encompass_Preferred
Human PLG (Plasminogen) ELISA Kit for detecting PLG in plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 78.125-5000pg/ml. Sensitivity: 46.875pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P00747. Target Synonyms: PLG; plasminogen; Plg; plasmin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5723