Learn More
Biomatik Corporation Mouse Sdc2 (Syndecan-2) ELISA Kit
Supplier:  Biomatik Corporation EKF58376
Encompass_Preferred
Mouse Sdc2 (Syndecan-2) ELISA Kit for detecting Sdc2 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.313-20ng/ml. Sensitivity: 0.188ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P43407. Target Synonyms: Sdc2; HSPG; CD362; Fibroglycan; HSPG1; SDC2; SYND2; CD362 antigen; Fibroglycan; fibroglycan; heparan sulfate proteoglycan 1; cell surface-associated; Heparan sulfate proteoglycan core protein; HSPG; HSPG1syndecan proteoglycan 2; SYND2cell surface-associated heparan sulfate proteoglycan 1; syndecan 2; syndecan-2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5736