Learn More
Biomatik Corporation 6-keto-PGF1a (6-keto-prostaglandin F1a) ELISA Kit
Supplier:  Biomatik Corporation EKF58381
Encompass_Preferred
6-keto-PGF1a (6-keto-prostaglandin F1a) ELISA Kit for detecting 6-keto-PGF1a in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Universal. Detection Range: 15.625-1000pg/ml. Sensitivity: 9.375pg/ml. Format: 96-Well Plate. Assay Type: Competitive ELISA, Coated with Antigen. Assay Time: 3h, 30 min. Target Synonyms: 6-keto-PGF1a; 6-keto-prostaglandin F1a. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5740