Learn More
Biomatik Corporation Rat IL-2Rα (Interleukin-2 Receptor-α) ELISA Kit
Supplier:  Biomatik Corporation EKF58394
Encompass_Preferred
Rat IL-2Rα (Interleukin-2 Receptor-α) ELISA Kit for detecting IL-2Rα in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.313-20ng/ml. Sensitivity: 0.188ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P26897. Target Synonyms: IL-2Rα; CD25; Interleukin 2 Receptor Alpha. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5749