Learn More
Biomatik Corporation Mouse Slc40a1 (Solute carrier family 40 member 1) ELISA Kit
Supplier:  Biomatik Corporation EKF58399
Encompass_Preferred
Mouse Slc40a1 (Solute carrier family 40 member 1) ELISA Kit for detecting Slc40a1 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.313-20ng/ml. Sensitivity: 0.188ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q9JHI9. Target Synonyms: Slc40a1; FPN1; HFE4; IREG1; MST079; MTP1; SLC11A3; SLC40A1; Ferroportin-1; FPN1IREG1ferroportin 1; HFE4ferroportin-1; Iron-regulated transporter 1; member 3; MST079; MTP1; putative ferroportin 1 variant IIIB; SLC11A3iron regulated gene 1; solute carrier family 11; proton-coupled divalent metal ion transporters; solute carrier family 40; iron-regulated transporter; member 1; solute carrier family 40 member 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5752