Learn More
Biomatik Corporation Rat Trpv1 (Transient receptor potential cation channel subfamily V member 1) ELISA Kit
Supplier:  Biomatik Corporation EKF58421
Encompass_Preferred
Rat Trpv1 (Transient receptor potential cation channel subfamily V member 1) ELISA Kit for detecting Trpv1 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 78.125-5000pg/ml. Sensitivity: 46.875pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: O35433. Target Synonyms: Trpv1; Capsaicin Receptor; TRPV1; VR1; Capsaicin receptor; Osm-9-like TRP channel 1; OTRPC1; transient receptor potential cation channel subfamily V member 1; transient receptor potential cation channel; subfamily V; member 1; transient receptor potential vanilloid 1a; transient receptor potential vanilloid 1b; TrpV1; Vanilloid receptor 1; vanilloid receptor subtype 1; VR1; VR1DKFZp434K0220. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5765