Learn More
Biomatik Corporation Mouse FE (Ferritin) ELISA Kit
Supplier:  Biomatik Corporation EKF58431
Encompass_Preferred
Mouse FE (Ferritin) ELISA Kit for detecting FE in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.469-30ng/ml. Sensitivity: 0.281ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: A2AHC6. Target Synonyms: Ferritin; FE. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5771