Learn More
Biomatik Corporation Rat Jag1 (Protein jagged-1) ELISA Kit
Supplier:  Biomatik Corporation EKF58438
Encompass_Preferred
Rat Jag1 (Protein jagged-1) ELISA Kit for detecting Jag1 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 78.125-5000pg/ml. Sensitivity: 46.875pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q63722. Target Synonyms: Jag1; CD339; AGS; AHDMGC104644; Alagille syndrome; AWS; CD339; CD339 antigen; HJ1; hJ1; jagged 1; Jagged1; JAGL1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5776