Learn More
Biomatik Corporation Rat IgG (Immunoglobulin G) ELISA Kit
Supplier:  Biomatik Corporation EKF58445
Encompass_Preferred
Rat IgG (Immunoglobulin G) ELISA Kit for detecting IgG in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 1.563-100ng/ml. Sensitivity: 0.938ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P20759; P20760. Target Synonyms: IgG; Immunoglobulin G. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5781