Learn More
Biomatik Corporation Mouse Wnt3a (Protein Wnt-3a) ELISA Kit
Supplier:  Biomatik Corporation EKF58447
Encompass_Preferred
Mouse Wnt3a (Protein Wnt-3a) ELISA Kit for detecting Wnt3a in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.313-20ng/ml. Sensitivity: 0.188ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P27467. Target Synonyms: Wnt3a; WNT3A; MGC119418; MGC119419; MGC119420; protein Wnt-3a; wingless-type MMTV integRation site family; member 3A. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5783