Learn More
Biomatik Corporation Mouse TLR4 (Toll-like receptor 4) ELISA Kit
Supplier:  Biomatik Corporation EKF58462
Encompass_Preferred
Mouse TLR4 (Toll-like receptor 4) ELISA Kit for detecting TLR4 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.313-20ng/ml. Sensitivity: 0.188ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q9QUK6. Target Synonyms: TLR4; CD284; ARMD10; CD284 antigen; Lps; hTollhomolog of Drosophila toll; TOLL; toll-like receptor 4. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5794