Learn More
Biomatik Corporation Rat Cys-C (Cystatin C) ELISA Kit
Supplier:  Biomatik Corporation EKF58465
Encompass_Preferred
Rat Cys-C (Cystatin C) ELISA Kit for detecting Cys-C in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 15.625-1000pg/ml. Sensitivity: 9.375pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P14841. Target Synonyms: Cystatin C; Cystatin-C; CST3; Cys-C; Cystatin 3; ARMD11; Post-Gamma-Globulin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5796