Learn More
Biomatik Corporation Rat G-6-Pase (Glucose-6-phosphatase) ELISA Kit
Supplier:  Biomatik Corporation EKF58476
Encompass_Preferred
Rat G-6-Pase (Glucose-6-phosphatase) ELISA Kit for detecting G-6-Pase; G6PT; GSD in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 78.125-5000miu/ml. Sensitivity: 46.875miu/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P43428. Target Synonyms: G-6-Pase; G6PT; GSD. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5805