Learn More
Biomatik Corporation Human SYNPO (Synaptopodin) ELISA Kit
Supplier:  Biomatik Corporation EKF58499
Encompass_Preferred
Human SYNPO (Synaptopodin) ELISA Kit for detecting SYNPO in cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q8N3V7. Target Synonyms: SYNPO; KIAA1029; Synaptopodin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5819