Learn More
Biomatik Corporation Human IHH (Indian hedgehog protein) ELISA Kit
Supplier:  Biomatik Corporation EKF58502
Encompass_Preferred
Human IHH (Indian hedgehog protein) ELISA Kit for detecting IHH in cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q14623. Target Synonyms: IHH; HHG2; HHG-2; BDA1; Ihh; Indian hedgehog homolog; Drosophila; Indian hedgehog; Indian hedgehog; Drosophila homolog; indian hedgehog protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5822