Learn More
Biomatik Corporation Human PDE5A (cGMP-specific 3',5'-cyclic phosphodiesterase) ELISA Kit
Supplier:  Biomatik Corporation EKF58519
Encompass_Preferred
Human PDE5A (cGMP-specific 3',5'-cyclic phosphodiesterase) ELISA Kit for detecting PDE5A in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: O76074. Target Synonyms: PDE5A; cGMP-specific 3'; 5'-cyclic phosphodiesterase; ; cGMP-binding cGMP-specific phosphodiesterase; CGB-PDE; PDE5. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5835