Learn More
Biomatik Corporation Rat Pon1 (Serum paraoxonase/arylesterase 1) ELISA Kit
Supplier:  Biomatik Corporation EKF58532
Encompass_Preferred
Rat Pon1 (Serum paraoxonase/arylesterase 1) ELISA Kit for detecting Pon1 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.781-50ng/ml. Sensitivity: 0.469ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P55159. Target Synonyms: Pon1; ESA; K-45; A-esterase 1; Aromatic esterase 1; arylesterase B-type; ESAesterase A; K-45; MVCD5; paraoxonase 1; PON 1; PONparaoxonase B-type; serum aryldiakylphosphatase; Serum aryldialkylphosphatase 1; serum paraoxonase; arylesterase 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5846