Learn More
Biomatik Corporation Rat α-GST (α-Glutathione S-Transferases) ELISA Kit
Supplier:  Biomatik Corporation EKF58534
Encompass_Preferred
Rat α-GST (α-Glutathione S-Transferases) ELISA Kit for detecting α-GST in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 37.5-2400pg/ml. Sensitivity: 22.5pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P00502. Target Synonyms: α-GST. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5848