Learn More
Biomatik Corporation Human LGMN (Legumain) ELISA Kit
Supplier:  Biomatik Corporation EKF58543
Encompass_Preferred
Human LGMN (Legumain) ELISA Kit for detecting LGMN in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q99538. Target Synonyms: LGMN; Legumain; AEP; LGMN1; PRSC1; Protease; cysteine 1; Asparaginyl Endopeptidase; cysteine protease 1; legumain; protease; cysteine; 1; legumain. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5856