Learn More
Biomatik Corporation Mouse CA9 (Carbonic anhydrase 9) ELISA Kit
Supplier:  Biomatik Corporation EKF58548
Encompass_Preferred
Mouse CA9 (Carbonic anhydrase 9) ELISA Kit for detecting CA9 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.625-1000pg/ml. Sensitivity: 9.375pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q8VHB5. Target Synonyms: CA9; ; Carbonic anhydrase 9; CAIX; CA-IX; MN; Carbonic DehydRatase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5859