Learn More
Biomatik Corporation Human FOS (Proto-oncogene c-Fos) ELISA Kit
Supplier:  Biomatik Corporation EKF58552
Encompass_Preferred
Human FOS (Proto-oncogene c-Fos) ELISA Kit for detecting FOS in cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.313-20ng/ml. Sensitivity: 0.188ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P01100. Target Synonyms: FOS; Proto-oncogene c-Fos; Cellular oncogene fos; G0; G1 switch regulatory protein 7; G0S7. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5863