Learn More
Biomatik Corporation Rat CK-18/KRT18 (CytokeRatin 18) ELISA Kit
Supplier:  Biomatik Corporation EKF58573
Encompass_Preferred
Rat CK-18/KRT18 (CytokeRatin 18) ELISA Kit for detecting CK-18; KRT18 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q5BJY9. Target Synonyms: KRT18; CK-18; CytokeRatin 18; KeRatin; type I cytoskeletal 18; KeRatin-18; K18; Cell prolifeRation-inducing gene 46 protein; CytokeRatin-18. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5878