Learn More
Biomatik Corporation Mouse Klk1 (Kallikrein-1) ELISA Kit
Supplier:  Biomatik Corporation EKF58588
Encompass_Preferred
Mouse Klk1 (Kallikrein-1) ELISA Kit for detecting Klk1 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P15947. Target Synonyms: Klk1; Kallikrein-1; Kidney; pancreas; salivary gland kallikrein; Tissue kallikrein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5889