Learn More
Biomatik Corporation Mouse ABCG1 (ATP Binding Cassette Transporter G1) ELISA Kit
Supplier:  Biomatik Corporation EKF58595
Encompass_Preferred
Mouse ABCG1 (ATP Binding Cassette Transporter G1) ELISA Kit for detecting ABCG1 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q64343. Target Synonyms: ABCG1; ATP-binding cassette sub-family G member 1; ATP-binding cassette transporter 8; White protein homolog. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5895